Anti-CD74 Mouse Monoclonal Antibody [clone: [LN2]] (Biotin)

49-209
10078-680EA 540.97 USD
10078-680
Anti-CD74 Mouse Monoclonal Antibody [clone: [LN2]] (Biotin)
CD74 antibody can be used in ELISA starting at 1:5000 - 1:40000, Western Blot starting at 1:5000 - 1:40000, and immunohistochemistry starting at 5 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store CD74 antibody undiluted at 4 °C.

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: [LN2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Baboon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR