Anti-BSG Mouse Monoclonal Antibody [clone: [HIM6]]

Supplier: ProSci
49-195
10078-660EA 684.46 USD
10078-660
Anti-BSG Mouse Monoclonal Antibody [clone: [HIM6]]
Antibodies
  • Basigin antibody can be used in immunohistochemistry starting at 10 μg/mL, and immunohistochemistry in frozen tissues starting at 1:400 - 1:600.

Affinity Chromatography

PBS, pH 7.2, 0.09% sodium azide.

Store Basigin antibody undiluted at 4 °C.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: [HIM6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR