Anti-GCG Rabbit Polyclonal Antibody

Supplier: Prosci

49-163
10076-256EA 647.66 USD
10076-256
Anti-GCG Rabbit Polyclonal Antibody
Antibodies
Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. Human, rabbit, rat, pig, and cow glucagons are identical. Glucagon is a member of a multigene family that includes secretim, vasoactive intestinal peptide gastric inhibitory peptide, glicentin, and others.

Glucagon antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Affinity Chromatography

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR