Anti-SOX2 Rabbit Polyclonal Antibody

Supplier: Prosci

49-142
10078-632EA 582.9 USD
10078-632
Anti-SOX2 Rabbit Polyclonal Antibody
Antibodies
SOX2 antibody can be used in ELISA, Western Blot starting at 1:5000 - 1:20000, and immunohistochemistry starting at 1:50.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR