Retinoic acid receptor alpha (RAR alpha), a NR1 Thyroid Hormone-Like Receptor, has a key effect on the development of acute promyelocytic leukemia (APL). APL results from chromosomal translocation of RAR alpha to various partner genes, including the promyelocytic leukemia (PML) gene on 15q22, the promyelocytic leukemia zinc finger (PLZF) gene on 11q23, the nucleophosmin (NPM) gene on 5q35, the nuclear mitotic apparatus (NuMA) gene on 11q13, and the signal transducer and activator of transcription STAT5b. The aberrant RAR alpha fusion proteins contribute to leukemic transformation by dominant inhibition of the expression of target genes that are important for cellular differentiation. Four alternatively spliced isoforms of RAR alpha have been documented in humans (RAR alpha1, RAR alpha2, RAR alpha1DeltaBC, and RAR alpha1DeltaB), one of which, RAR alpha1DeltaB, does not code for a functional receptor.
Retinoic Acid Receptor alpha antibody can be used in ELISA starting at 1:100 - 1:500, Western Blot, and immunohistochemistry starting at 10 ?g/mL.
Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human