Anti-PTK2B Rabbit Polyclonal Antibody

Supplier: ProSci
49-131
10078-620EA 674.53 USD
10078-620
Anti-PTK2B Rabbit Polyclonal Antibody
Antibodies
Focal adhesion kinase 2, also known as proline-rich tyrosine kinase 2 (PYK2), cell adhesion kinase, related adhesion focal tyrosine kinase, and calcium-dependent protein tyrosine kinase, has been implicated in multiple signaling events, including modulation of ion channels, T- and B-cell receptor signaling, and cell death.

PTK2B antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1:10 - 1:100.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTK2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR