Anti-NCAM1 Mouse Monoclonal Antibody [clone: [HCD56]]

Supplier: Prosci

49-120
10078-604EA 582.74 USD
10078-604
Anti-NCAM1 Mouse Monoclonal Antibody [clone: [HCD56]]
Antibodies
NCAM1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide. The.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: [HCD56]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR