Anti-SYK Mouse Monoclonal Antibody [clone: [SYK-01]]

Supplier: ProSci
49-116
10076-252EA 674.53 USD
10076-252
Anti-SYK Mouse Monoclonal Antibody [clone: [SYK-01]]
Antibodies
SYK antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone: [SYK-01]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR