Clusterin (SGP-2, SP-40,40, pADHC-9, TRPM-2, CLJ, T64, GP III, APO J, XIP8) is a 75-80 kD disulfide-linked heterodimeric protein containing about 30% of N-linked carbohydrate rich in sialic acid but truncated forms targeted to the nucleus have also been identified. It is a conserved secreted glycoprotein expressed by a wide range of tissues and being implicated in several physiological processes. Across a broad range of species clusterin shows a high degree of sequence homology ranging from 70% to 80%. It is nearly ubiqitously expressed in most mammalian tissues and can be found in plasma, milk, urine, cerebrospinal fluid and semen. It is able to bind and form complexes with numerous partners such as immunoglobulins, lipids, heparin, bacteria, complement components, paraoxonase, beta amyloid, leptin and others. Clusterin has a number of functions such as phagocyte recruitment, aggregation induction, complement attack prevention, apoptosis inhibition, membrane remodelling, lipid transport, hormone transport and/or scavenging, matrix metalloproteinase inhibition. One tempting hypothesis says that clusterin is an extracellular chaperone protecting cells from stress-induced insults caused by degraded and misfolded protein precipitates. The protein is expressed in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others. A genuine function of clusterin has not been defined.
Clusterin antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.
Protein A Column
PBS, 15 mM sodium azide, approx., pH 7.4.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CLU
Clonality: Monoclonal
Clone: [Hs-3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human