Anti-TUBA1B Mouse Monoclonal Antibody [clone: [TU-01]]

Supplier: Prosci

49-105
10076-230EA 561.89 USD
10076-230
Anti-TUBA1B Mouse Monoclonal Antibody [clone: [TU-01]]
Antibodies
TUBA antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUBA1B
Clonality: Monoclonal
Clone: [TU-01]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR