Anti-TF Mouse Monoclonal Antibody [clone: [2A2]]

Supplier: ProSci
49-098
10076-216EA 698.16 USD
10076-216
Anti-TF Mouse Monoclonal Antibody [clone: [2A2]]
Antibodies
Transferrin antibody can be used in immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone: [2A2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR