Anti-DIABLO Mouse Monoclonal Antibody [clone: [79-1-83]]

Supplier: ProSci
49-090
10076-200EA 674.53 USD
10076-200
Anti-DIABLO Mouse Monoclonal Antibody [clone: [79-1-83]]
Antibodies
SMAC antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 ?g/mL.

PBS, pH 7.2, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DIABLO
Clonality: Monoclonal
Clone: [79-1-83]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR