Anti-PDGFRB Mouse Monoclonal Antibody [clone: [42G12]]

Supplier: ProSci
49-089
10076-198EA 698.16 USD
10076-198
Anti-PDGFRB Mouse Monoclonal Antibody [clone: [42G12]]
Antibodies
PDGFR antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry starting at 1:10 - 1:50.

PBS, pH 7.2, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: [42G12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR