Anti-ALPL Mouse Monoclonal Antibody

Supplier: ProSci
49-088
10078-596EA 698.16 USD
10078-596
Anti-ALPL Mouse Monoclonal Antibody
Antibodies
ALPL antibody can be used in Western Blot, and immunohistochemistry starting at 1 - 2 ?g/mL.

PBS, pH 7.2, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR