Anti-CALR Mouse Monoclonal Antibody [clone: [1G6A7]]

Supplier: ProSci
49-068
10078-556EA 684.46 USD
10078-556
Anti-CALR Mouse Monoclonal Antibody [clone: [1G6A7]]
Antibodies
Calreticulin antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 5 ?g/mL, immunofluorescence, and immunoprecipitation.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CALR
Clonality: Monoclonal
Clone: [1G6A7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR