Anti-LPP Mouse Monoclonal Antibody

Supplier: Prosci

49-067
10078-554EA 582.9 USD
10078-554
Anti-LPP Mouse Monoclonal Antibody
Antibodies
LPP antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:500, and immunofluorescence.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR