Anti-KLK3 Mouse Monoclonal Antibody [clone: [5A11E2]]

Supplier: Prosci

49-065
10078-550EA 582.9 USD
10078-550
Anti-KLK3 Mouse Monoclonal Antibody [clone: [5A11E2]]
Antibodies
PSA antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: [5A11E2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR