Anti-STYK1 Mouse Monoclonal Antibody [clone: [2H2F10]]

Supplier: Prosci

49-064
10078-548EA 582.9 USD
10078-548
Anti-STYK1 Mouse Monoclonal Antibody [clone: [2H2F10]]
Antibodies
STYK1 antibody can be used in ELISA starting at 1:10000 - 1:44000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STYK1
Clonality: Monoclonal
Clone: [2H2F10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR