Anti-DDR2 Mouse Monoclonal Antibody

Supplier: Prosci

49-061
10078-542EA 582.9 USD
10078-542
Anti-DDR2 Mouse Monoclonal Antibody
Antibodies
DDR2 antibody can be used in ELISA, and immunohistochemistry starting at 2.5 mg/ml.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDR2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR