Anti-CK5 Mouse Monoclonal Antibody

Supplier: Prosci

49-058
10078-536EA 582.9 USD
10078-536
Anti-CK5 Mouse Monoclonal Antibody
Antibodies
Cytoke antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR