Anti-FGFR4 Mouse Monoclonal Antibody [clone: [4H2B10B2]]

Supplier: ProSci
49-057
10078-534EA 674.53 USD
10078-534
Anti-FGFR4 Mouse Monoclonal Antibody [clone: [4H2B10B2]]
Antibodies
FGFR4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 ?g/mL.

0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FGFR4
Clonality: Monoclonal
Clone: [4H2B10B2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR