Anti-PECAM1 Mouse Monoclonal Antibody

Supplier: ProSci
49-056
10076-196EA 674.53 USD
10076-196
Anti-PECAM1 Mouse Monoclonal Antibody
Antibodies
CD31 antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:1000 - 1:5000, and immunohistochemistry starting at 2.5 ?g/mL.

0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR