Anti-TUBB Mouse Monoclonal Antibody

Supplier: Prosci

49-054
10076-192EA 582.9 USD
10076-192
Anti-TUBB Mouse Monoclonal Antibody
Antibodies
Tubulin Beta antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide, before the addition of glycerol to 40%.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUBB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Bovine, Chicken, Frog, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR