Anti-HDAC1 Mouse Monoclonal Antibody

Supplier: Prosci

49-052
10076-188EA 582.9 USD
10076-188
Anti-HDAC1 Mouse Monoclonal Antibody
Antibodies
  • HDAC1 antibody can be used in immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HDAC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Gorilla, Hamster, Human, Monkey, Mouse, Orangutan, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR