Anti-PINK1 Mouse Monoclonal Antibody

49-049
10076-182EA 491.23 USD
10076-182
Anti-PINK1 Mouse Monoclonal Antibody
PINK1 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PINK1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR