Anti-SOCS3 Mouse Monoclonal Antibody [clone: [SO1]]

Supplier: ProSci

49-045
10076-174EA 630.41 USD
10076-174
Anti-SOCS3 Mouse Monoclonal Antibody [clone: [SO1]]
Antibodies
SOCS3 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 2.5 ?g/mL, and immunoprecipitation

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: [SO1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR