Anti-BAD Mouse Monoclonal Antibody [clone: [BYC001]]

49-038
10078-522EA 540.97 USD
10078-522
Anti-BAD Mouse Monoclonal Antibody [clone: [BYC001]]
BAD antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BAD
Clonality: Monoclonal
Clone: [BYC001]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR