Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOD1 Mouse Monoclonal Antibody

Supplier: Prosci

49-037
10078-520EA 515.21 USD InStock
10078-520
Anti-SOD1 Mouse Monoclonal Antibody
Antibodies
SOD1 antibody can be used in immunocytochemistry starting at 1:50, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 1:100.

Protein G Column

PBS, 0.09% sodium azide, 0.1% BSA.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SOD1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty