Anti-ZAP70 Mouse Monoclonal Antibody [clone: [11H40]]

Supplier: ProSci
49-036
10078-518EA 684.46 USD
10078-518
Anti-ZAP70 Mouse Monoclonal Antibody [clone: [11H40]]
Antibodies
ZAP70 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 0.09% azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: [11H40]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR