Anti-TG Mouse Monoclonal Antibody

Supplier: ProSci
49-029
10078-504EA 540.97 USD
10078-504
Anti-TG Mouse Monoclonal Antibody
Thyroglobulin antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide, 40% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR