Anti-TGM4 Mouse Monoclonal Antibody

49-009
10076-172EA 540.97 USD
10076-172
Anti-TGM4 Mouse Monoclonal Antibody
Antibodies
Transglutaminase 4 antibody can be used in Western Blot starting at 5 - 3 ?g/mL, immunohistochemistry starting at 10 ?g/mL, and flow cytometry.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TGM4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR