Anti-ST13 Mouse Monoclonal Antibody

49-007
10076-168EA 491.23 USD
10076-168
Anti-ST13 Mouse Monoclonal Antibody
Himmunoprecipitation antibody can be used in ELISA starting at 1:2000 - 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ST13
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR