Anti-CRABP2 Mouse Monoclonal Antibody [clone: [AT2E11]]

Supplier: ProSci
49-005
10076-164EA 674.53 USD
10076-164
Anti-CRABP2 Mouse Monoclonal Antibody [clone: [AT2E11]]
Antibodies
CRABP2 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRABP2
Clonality: Monoclonal
Clone: [AT2E11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR