Anti-CD34 Mouse Monoclonal Antibody [clone: [4H11[APG]]]

48-996
10076-150EA 540.97 USD
10076-150
Anti-CD34 Mouse Monoclonal Antibody [clone: [4H11[APG]]]
The antibody 4H11[APG] reacts with Class III epitope on CD34 (Mucosialin), a 110-115 kDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells.

CD34 antibody can be used in ELISA, Western Blot starting at 1:250 - 1:2500, and immunohistochemistry starting at 5 ?g/mL.

Precipitation

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: [4H11[APG]]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR