Anti-CD59 Mouse Monoclonal Antibody [clone: [MEM-43]]

48-991
10078-468EA 540.97 USD
10078-468
Anti-CD59 Mouse Monoclonal Antibody [clone: [MEM-43]]
CD59 antibody can be used in Western Blot starting at 1:1000, immunohistochemistry starting at 1:1000, immunohistochemistry in frozen tissues starting at 1:1000, and immunofluorescence starting at 1:1000.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: [MEM-43]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR