Anti-CD55 Mouse Monoclonal Antibody [clone: [MEM-118]]

48-990
10078-466EA 674.53 USD
10078-466
Anti-CD55 Mouse Monoclonal Antibody [clone: [MEM-118]]
Antibodies
CD55 is widely expressed on hematopoietic and on many non-hematopoietic cells; it is weakly present on NK cells. Decay-accelerating Factor (CD55) is one of the three proteins protecting cells from autologous lysis by complement. (The others are: membrane cofactor protein (MCP; CD46) and an inhibitor of the membrane attack complex formation (CD59))

CD55 antibody can be used in Western Blot starting at 1.5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

Size Exclusion Chromatography

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: [MEM-118]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR