Anti-CD5 Mouse Monoclonal Antibody [clone: [MEM-32]]

48-984
10078-454EA 540.97 USD
10078-454
Anti-CD5 Mouse Monoclonal Antibody [clone: [MEM-32]]
CD5 antibody can be used in Western Blot starting at 1:1000 - 1:10000, and immunohistochemistry starting at 1:100.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: [MEM-32]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR