Anti-TOM1L1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-981
10078-448EA 582.9 USD
10078-448
Anti-TOM1L1 Rabbit Polyclonal Antibody
Antibodies
TOM1L1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TOM1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR