Anti-SLC4A1 Rabbit Polyclonal Antibody

Supplier: Prosci
48-976
10078-438EA 785.68 USD
10078-438
Anti-SLC4A1 Rabbit Polyclonal Antibody
Antibodies
SLC4A1 antibody can be used in Western Blot, immunohistochemistry starting at 1:50, and immunoprecipitation.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR