Anti-SLC15A1 Rabbit Polyclonal Antibody

Supplier: Prosci
48-974
10078-434EA 795.23 USD
10078-434
Anti-SLC15A1 Rabbit Polyclonal Antibody
Antibodies
SLC15A1 antibody can be used in Western Blot starting at 5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC15A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR