Anti-NOV Rabbit Polyclonal Antibody

Supplier: Prosci

48-970
10078-426EA 582.9 USD
10078-426
Anti-NOV Rabbit Polyclonal Antibody
Antibodies
NOV antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR