RDC1 was originally considered to be an Adrenomedullin receptor that binds calcitonin gene-related peptide (CGRP) and adrenomedullin. RDC1 and GPRN1 represent two tissue-specific subtypes of the same 60- kDa receptor. RDC1 has the structural characteristics of a chemokine receptor. Recently it has been shown that RDC1 binds to the chemokine CXCL12 and should be considered to be a chemokine receptor.
CXCR7 antibody can be used in immunohistochemistry, Western Blot starting at 1:500, and immunohistochemistry starting at 1:100.
Protein G Column
PBS, 0.05% BSA, 0.05% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CXCR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat