Anti-FOXO3 Rabbit Polyclonal Antibody

Supplier: Prosci

48-953
10078-392EA 582.9 USD
10078-392
Anti-FOXO3 Rabbit Polyclonal Antibody
Antibodies
FOXO3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Xenopus, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR