Anti-HAPLN3 Rabbit Polyclonal Antibody

48-941
10076-144EA 674.53 USD
10076-144
Anti-HAPLN3 Rabbit Polyclonal Antibody
Antibodies
HAPLN3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HAPLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR