Anti-PPP1R1A Rabbit Polyclonal Antibody

48-938
10076-138EA 698.16 USD
10076-138
Anti-PPP1R1A Rabbit Polyclonal Antibody
Antibodies
PPP1R1A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 - 20 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPP1R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Xenopus, Horse, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR