Anti-HCAR3 Rabbit Polyclonal Antibody

Supplier: Prosci

48-921
10076-104EA 582.9 USD
10076-104
Anti-HCAR3 Rabbit Polyclonal Antibody
Antibodies
GPR109B/HM74 is a Nicotinic Acid GPCR.

GPR109B antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HCAR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Dog, Hamster, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR