Anti-GPR77 Rabbit Polyclonal Antibody

Supplier: Prosci

48-918
10076-098EA 582.9 USD
10076-098
Anti-GPR77 Rabbit Polyclonal Antibody
Antibodies
C5L2 is an Anaphylatoxin Receptor. Binding to anaphylatoxins can result in increased triglyceride synthesis, although its precise role in cellular responses has not been determined.

GPR77 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR