Anti-KDM1A Rabbit Polyclonal Antibody

Supplier: Prosci

48-917
10076-096EA 582.9 USD
10076-096
Anti-KDM1A Rabbit Polyclonal Antibody
Antibodies
AOF2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KDM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR