Anti-ZMYM2 Rabbit Polyclonal Antibody

Supplier: Prosci
48-915
10076-092EA 785.68 USD
10076-092
Anti-ZMYM2 Rabbit Polyclonal Antibody
Antibodies
ZMYM2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZMYM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Human, Monkey, Mouse, Orangutan, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR