Anti-MCTS1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-875
10076-072EA 582.9 USD
10076-072
Anti-MCTS1 Rabbit Polyclonal Antibody
Antibodies
MCTS1 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MCTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Bovine, Chicken, Dog, Xenopus, Human, Mouse, Pig, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR